accessibility menu, dialog, popup

UserName

CAS RN 2958-36-3

IUPAC Name: 4-chloro-2-(2-chlorobenzoyl)aniline
Synonyms: 4-chloro-2-(2-chlorobenzoyl)aniline
Molecular Weight (g/mol): 266.12
Molecular Formula: C13H9Cl2NO
InChi Key: KWZYIAJRFJVQDO-UHFFFAOYSA-N
SMILES: NC1=CC=C(Cl)C=C1C(=O)C1=CC=CC=C1Cl
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)