missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-2',5-dichlorobenzophenone, 98%

CAS: 2958-36-3 | C13H9Cl2NO | 266.121 g/mol

Supplier:  Thermo Scientific Chemicals B2036018

 View more versions of this product

Catalog No. AAB2036018


Only null left
Explore available promotions

Description

Description

2-Amino-2',5-dichlorobenzophenone is used to produce lorazepam a drug of the benzodiazepine group, used especially to treat anxiety.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-2′,5-dichlorobenzophenone is used to produce lorazepam a drug of the benzodiazepine group, used especially to treat anxiety.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

2-Amino-2',5-dichlorobenzophenone
87°C to 89°C
C13H9Cl2NO
50 g
2-amino-2',5-dichlorobenzophenone, 2-amino-5-chlorophenyl 2-chlorophenyl methanone, 4-chloro-2-2-chlorobenzoyl aniline, methanone, 2-amino-5-chlorophenyl 2-chlorophenyl, unii-g806veo3ke, 2-amino-5-chloro-2'-chlorobenzophenone, 2-amino-5-chlorophenyl-2-chlorophenyl methanone, g806veo3ke, benzophenone, 2-amino-2',5-dichloro, 2-amino-5,2'-dichlorobenzophenone
KWZYIAJRFJVQDO-UHFFFAOYSA-N
(2-amino-5-chlorophenyl)-(2-chlorophenyl)methanone
18069
99%
2958-36-3
Odorless
MFCD00007840
652774
Insoluble in water.
C1=CC=C(C(=C1)C(=O)C2=C(C=CC(=C2)Cl)N)Cl
266.121
266.13

Chemical Identifiers

2958-36-3
266.121
KWZYIAJRFJVQDO-UHFFFAOYSA-N
18069
C1=CC=C(C(=C1)C(=O)C2=C(C=CC(=C2)Cl)N)Cl
C13H9Cl2NO
MFCD00007840
2-amino-2',5-dichlorobenzophenone, 2-amino-5-chlorophenyl 2-chlorophenyl methanone, 4-chloro-2-2-chlorobenzoyl aniline, methanone, 2-amino-5-chlorophenyl 2-chlorophenyl, unii-g806veo3ke, 2-amino-5-chloro-2'-chlorobenzophenone, 2-amino-5-chlorophenyl-2-chlorophenyl methanone, g806veo3ke, benzophenone, 2-amino-2',5-dichloro, 2-amino-5,2'-dichlorobenzophenone
(2-amino-5-chlorophenyl)-(2-chlorophenyl)methanone
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

EINECSNumber : 220-985-2

RTECSNumber : DJ0200000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only