missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-2',5-dichlorobenzophenone, 98%

Catalog No. AAB2036006
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2036006 5 g
AAB2036009 10 g
AAB2036018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2036006 Supplier Thermo Scientific Chemicals Supplier No. B2036006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2958-36-3 | C13H9Cl2NO | 266.121 g/mol

2-Amino-2',5-dichlorobenzophenone is used to produce lorazepam a drug of the benzodiazepine group, used especially to treat anxiety.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-2′,5-dichlorobenzophenone is used to produce lorazepam a drug of the benzodiazepine group, used especially to treat anxiety.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 2958-36-3
Molecular Formula C13H9Cl2NO
Molecular Weight (g/mol) 266.121
MDL Number MFCD00007840
InChI Key KWZYIAJRFJVQDO-UHFFFAOYSA-N
Synonym 2-amino-2',5-dichlorobenzophenone, 2-amino-5-chlorophenyl 2-chlorophenyl methanone, 4-chloro-2-2-chlorobenzoyl aniline, methanone, 2-amino-5-chlorophenyl 2-chlorophenyl, unii-g806veo3ke, 2-amino-5-chloro-2'-chlorobenzophenone, 2-amino-5-chlorophenyl-2-chlorophenyl methanone, g806veo3ke, benzophenone, 2-amino-2',5-dichloro, 2-amino-5,2'-dichlorobenzophenone
PubChem CID 18069
IUPAC Name (2-amino-5-chlorophenyl)-(2-chlorophenyl)methanone
SMILES C1=CC=C(C(=C1)C(=O)C2=C(C=CC(=C2)Cl)N)Cl

Specifications

Melting Point 87°C to 89°C
Odor Odorless
Quantity 5 g
Beilstein 652774
Solubility Information Insoluble in water.
Formula Weight 266.13
Percent Purity 99%
Chemical Name or Material 2-Amino-2',5-dichlorobenzophenone
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 220-985-2
RTECSNumber DJ0200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only