missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-2',4-dichlorodiphenyl Ether 98.0+%, TCI America™

Catalog No. A22245G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A22245G 5 g
A222425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A22245G Supplier TCI America Supplier No. A22245G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 56966-48-4
Molecular Formula C12H9Cl2NO
Molecular Weight (g/mol) 254.11
MDL Number MFCD00025218
InChI Key PHORTNLNXNZNET-UHFFFAOYSA-N
Synonym 5-Chloro-2-(2-chlorophenoxy)aniline
PubChem CID 92601
IUPAC Name 5-chloro-2-(2-chlorophenoxy)aniline
SMILES C1=CC=C(C(=C1)OC2=C(C=C(C=C2)Cl)N)Cl

Specifications

Boiling Point 210°C
Quantity 5 g
Formula Weight 254.11
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Amino-2′,4-dichlorodiphenyl Ether