accessibility menu, dialog, popup

UserName

CAS RN 56966-48-4

IUPAC Name: 5-chloro-2-(2-chlorophenoxy)aniline
Synonyms: 5-chloro-2-(2-chlorophenoxy)aniline
Molecular Weight (g/mol): 254.11
Molecular Formula: C12H9Cl2NO
InChi Key: PHORTNLNXNZNET-UHFFFAOYSA-N
SMILES: NC1=CC(Cl)=CC=C1OC1=CC=CC=C1Cl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results