missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-6-methoxynaphthalene 98.0+%, TCI America™

Catalog No. p-7117693
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A203025G 25 g
A2030100G 100 g
A2030500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A203025G Supplier TCI America Supplier No. A203025G
Only null left

Chemical Identifiers

CAS 3900-45-6
Molecular Formula C13H12O2
Molecular Weight (g/mol) 200.237
MDL Number MFCD00021643
InChI Key GGWCZBGAIGGTDA-UHFFFAOYSA-N
Synonym 2-acetyl-6-methoxynaphthalene, 1-6-methoxynaphthalen-2-yl ethanone, 1-6-methoxy-2-naphthyl ethanone, 6-methoxy-2-acetylnaphthalene, unii-61vbh3537c, 6'-methoxy-2'-acetonaphthone, 1-6-methoxy-2-naphthyl ethan-1-one, ethanone, 1-6-methoxy-2-naphthalenyl, 2-acetyl-6-methoxy naphthalene, 6-methoxy-2-acetyl naphthalene
PubChem CID 77506
IUPAC Name 1-(6-methoxynaphthalen-2-yl)ethanone
SMILES CC(=O)C1=CC2=C(C=C1)C=C(C=C2)OC

Specifications

Melting Point 109°C
Color Yellow
Boiling Point 195°C
Quantity 25 g
Formula Weight 200.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetyl-6-methoxynaphthalene