accessibility menu, dialog, popup

UserName

CAS RN 3900-45-6

IUPAC Name: 1-(6-methoxynaphthalen-2-yl)ethan-1-one
Synonyms: 1-(6-methoxynaphthalen-2-yl)ethanone
Molecular Weight (g/mol): 200.24
Molecular Formula: C13H12O2
InChi Key: GGWCZBGAIGGTDA-UHFFFAOYSA-N
SMILES: COC1=CC=C2C=C(C=CC2=C1)C(C)=O
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (3)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (5)
  • (2)
  • (3)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)