missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-6-methoxynaphthalene, 98%

CAS: 3900-45-6 | C13H12O2 | 200.237 g/mol

Supplier:  Thermo Scientific Chemicals A1803114

 View more versions of this product

Catalog No. AAA1803114

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Acetyl-6-methoxynaphthalene
107°C to 109°C
C13H12O2
25 g
2-acetyl-6-methoxynaphthalene, 1-6-methoxynaphthalen-2-yl ethanone, 1-6-methoxy-2-naphthyl ethanone, 6-methoxy-2-acetylnaphthalene, unii-61vbh3537c, 6'-methoxy-2'-acetonaphthone, 1-6-methoxy-2-naphthyl ethan-1-one, ethanone, 1-6-methoxy-2-naphthalenyl, 2-acetyl-6-methoxy naphthalene, 6-methoxy-2-acetyl naphthalene
CC(=O)C1=CC2=C(C=C1)C=C(C=C2)OC
200.237
200.24
3900-45-6
155°C to 160°C (0.4 mmHg)
MFCD00021643
1871098
GGWCZBGAIGGTDA-UHFFFAOYSA-N
1-(6-methoxynaphthalen-2-yl)ethanone
77506
98%

Chemical Identifiers

3900-45-6
200.237
GGWCZBGAIGGTDA-UHFFFAOYSA-N
77506
CC(=O)C1=CC2=C(C=C1)C=C(C=C2)OC
C13H12O2
MFCD00021643
2-acetyl-6-methoxynaphthalene, 1-6-methoxynaphthalen-2-yl ethanone, 1-6-methoxy-2-naphthyl ethanone, 6-methoxy-2-acetylnaphthalene, unii-61vbh3537c, 6'-methoxy-2'-acetonaphthone, 1-6-methoxy-2-naphthyl ethan-1-one, ethanone, 1-6-methoxy-2-naphthalenyl, 2-acetyl-6-methoxy naphthalene, 6-methoxy-2-acetyl naphthalene
1-(6-methoxynaphthalen-2-yl)ethanone
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 223-453-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only