missing translation for 'onlineSavingsMsg'
Learn More

2-(9-Anthryl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98.0+%, TCI America™

Catalog No. p-7118245
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A31841G 1 g
A31845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A31841G Supplier TCI America Supplier No. A31841G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 709022-63-9
Molecular Formula C20H21BO2
Molecular Weight (g/mol) 304.196
MDL Number MFCD24039741
InChI Key GBHRGHJTZFOAKE-UHFFFAOYSA-N
PubChem CID 18533854
IUPAC Name 2-anthracen-9-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C3C=CC=CC3=CC4=CC=CC=C24

Specifications

Melting Point 141°C
Color White-Yellow
Quantity 1 g
Formula Weight 304.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(9-Anthryl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane