accessibility menu, dialog, popup

UserName

BOOCH3CH3H3CCH3

CAS RN 709022-63-9

IUPAC Name: 2-(anthracen-9-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(anthracen-9-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 304.2
Molecular Formula: C20H21BO2
InChi Key: GBHRGHJTZFOAKE-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=C2C=CC=CC2=CC2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results