missing translation for 'onlineSavingsMsg'
Learn More

2-(9,9'-Spirobi[fluoren]-7-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98.0+%, TCI America™

Supplier:  TCI America S09805G

 View more versions of this product

Catalog No. S09805G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-(9,9′-Spirobi[fluoren]-7-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
250°C
C31H27BO2
(9,9′-Spirobi[fluoren]-7-yl)boronic Acid Pinacol Ester, 7-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9′-spirobi[fluorene]
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=CC=CC=C4C35C6=CC=CC=C6C7=CC=CC=C57
442.365
442.37
Crystalline Powder
884336-44-1
White-Yellow
5 g
VZMLUIPYIYNRGP-UHFFFAOYSA-N
4,4,5,5-tetramethyl-2-(9,9'-spirobi[fluorene]-2-yl)-1,3,2-dioxaborolane
60135725
≥98.0% (HPLC,T)

Chemical Identifiers

884336-44-1
442.365
60135725
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=CC=CC=C4C35C6=CC=CC=C6C7=CC=CC=C57
C31H27BO2
VZMLUIPYIYNRGP-UHFFFAOYSA-N
4,4,5,5-tetramethyl-2-(9,9'-spirobi[fluorene]-2-yl)-1,3,2-dioxaborolane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions