accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3C

CAS RN 884336-44-1

IUPAC Name: 4,4,5,5-tetramethyl-2-{9,9'-spirobi[fluoren]-2-yl}-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-{9,9'-spirobi[fluoren]-2-yl}-1,3,2-dioxaborolane
Molecular Weight (g/mol): 442.37
Molecular Formula: C31H27BO2
InChi Key: VZMLUIPYIYNRGP-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC2=C(C=C1)C1=CC=CC=C1C21C2=CC=CC=C2C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results