missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(9,9'-Spirobi[fluoren]-7-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98.0+%, TCI America™
$149.13 - $738.16
Chemical Identifiers
| CAS | 884336-44-1 |
|---|---|
| Molecular Formula | C31H27BO2 |
| Molecular Weight (g/mol) | 442.365 |
| InChI Key | VZMLUIPYIYNRGP-UHFFFAOYSA-N |
| PubChem CID | 60135725 |
| IUPAC Name | 4,4,5,5-tetramethyl-2-(9,9'-spirobi[fluorene]-2-yl)-1,3,2-dioxaborolane |
| SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=CC=CC=C4C35C6=CC=CC=C6C7=CC=CC=C57 |
Chemical Identifiers
| 884336-44-1 | |
| 442.365 | |
| 60135725 | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=CC=CC=C4C35C6=CC=CC=C6C7=CC=CC=C57 |
| C31H27BO2 | |
| VZMLUIPYIYNRGP-UHFFFAOYSA-N | |
| 4,4,5,5-tetramethyl-2-(9,9'-spirobi[fluorene]-2-yl)-1,3,2-dioxaborolane |
Specifications
| 884336-44-1 | |
| White-Yellow | |
| 1 g | |
| VZMLUIPYIYNRGP-UHFFFAOYSA-N | |
| 4,4,5,5-tetramethyl-2-(9,9'-spirobi[fluorene]-2-yl)-1,3,2-dioxaborolane | |
| 60135725 | |
| ≥98.0% (HPLC,T) | |
| 2-(9,9′-Spirobi[fluoren]-7-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
| 250°C | |
| C31H27BO2 | |
| (9,9′C-Spirobi[fluoren]-7-yl)boronic Acid Pinacol Ester, 7-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9′C-spirobi[fluorene] | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=CC=CC=C4C35C6=CC=CC=C6C7=CC=CC=C57 | |
| 442.365 | |
| 442.37 | |
| Crystalline Powder |
Safety and Handling
TSCA : No