missing translation for 'onlineSavingsMsg'
Learn More

2,7-Diiodofluorene 98.0+%, TCI America™

Catalog No. D45045G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45045G 5 g
D45041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45045G Supplier TCI America Supplier No. D45045G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16218-28-3
Molecular Formula C13H8I2
Molecular Weight (g/mol) 418.016
MDL Number MFCD00040153
InChI Key YCWGCTPMBCOCLT-UHFFFAOYSA-N
PubChem CID 634493
IUPAC Name 2,7-diiodo-9H-fluorene
SMILES C1C2=C(C=CC(=C2)I)C3=C1C=C(C=C3)I

Specifications

Melting Point 218°C
Color Yellow
Quantity 5 g
Formula Weight 418.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Diiodofluorene