accessibility menu, dialog, popup

UserName

II

CAS RN 16218-28-3

IUPAC Name: 2,7-diiodo-9H-fluorene
Synonyms: 2,7-diiodo-9H-fluorene
Molecular Weight (g/mol): 418.02
Molecular Formula: C13H8I2
InChi Key: YCWGCTPMBCOCLT-UHFFFAOYSA-N
SMILES: IC1=CC2=C(C=C1)C1=C(C2)C=C(I)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results