missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,7-Dihydroxynaphthalene 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | 186°C |
| Quantity | 100 g |
| Formula Weight | 160.17 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,7-Dihydroxynaphthalene |
Safety and Handling
| EINECSNumber | (4)-0356 |
| RTECSNumber | QJ4780000 |
| TSCA | Yes |