accessibility menu, dialog, popup

UserName

HOOH

CAS RN 582-17-2

IUPAC Name: naphthalene-2,7-diol
Synonyms: 2,7-naphthalenediol
Molecular Weight (g/mol): 160.17
Molecular Formula: C10H8O2
InChi Key: DFQICHCWIIJABH-UHFFFAOYSA-N
SMILES: OC1=CC2=CC(O)=CC=C2C=C1
Percent Purity 
  • (3)
  • (7)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (3)
  • (7)
  • (3)
  • (7)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (4)