missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2',7'-Dichlorofluorescein, ACS, Spectrum™ Chemical
$220.81 - $12414.41
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18608173
|
Spectrum Chemical Mfg Cor
DI1161GM |
1 g | Amber Glass Bottle |
Each for $220.81
|
|
||||
|
18608174
|
Spectrum Chemical Mfg Cor
DI1165GM |
5 g | Amber Glass Bottle |
Each for $582.41
|
|
||||
|
18608175
|
Spectrum Chemical Mfg Cor
DI11625GM |
25 g | Amber Glass Bottle |
Each for $1,736.84
|
|
||||
|
18608176
|
Spectrum Chemical Mfg Cor
DI116500GMBL |
500 g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 76-54-0 |
Specifications
| 76-54-0 | |
| ACS | |
| Brown | |
| 2',7'-Dichlorofluorescein |
| 1 | |
| Amber Glass Bottle | |
| 1 g |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.