missing translation for 'onlineSavingsMsg'
Learn More

2',7'-Dichlorofluorescein, ACS, Spectrum™ Chemical

Catalog No. 18608176
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
500 g
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608176 500 g Amber Glass Bottle
18608173 1 g Amber Glass Bottle
18608174 5 g Amber Glass Bottle
18608175 25 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18608176 Supplier Spectrum Chemical Mfg Cor Supplier No. DI116500GMBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 76-54-0 | 401.20 g/mol

Chemical Identifiers

CAS 76-54-0

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Color Brown
Quantity 500 g
Molecular Weight (g/mol) 401.20
Grade ACS
Chemical Name or Material 2',7'-Dichlorofluorescein

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.