missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,7-Dibromotriphenylene 98.0+%, TCI America™
$102.52 - $307.57
Chemical Identifiers
| CAS | 888041-37-0 |
|---|---|
| Molecular Formula | C18H10Br2 |
| Molecular Weight (g/mol) | 386.086 |
| MDL Number | MFCD22571695 |
| InChI Key | BPGPBYGXGRDFQG-UHFFFAOYSA-N |
| PubChem CID | 16122224 |
| IUPAC Name | 2,7-dibromotriphenylene |
| SMILES | C1=CC=C2C(=C1)C3=C(C=CC(=C3)Br)C4=C2C=C(C=C4)Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D4801200MG
|
TCI America
D4801200MG |
200 mg |
Each for $102.52
|
|
|||||
|
D48011G
|
TCI America
D48011G |
1 g |
Each for $307.57
|
|
|||||
Chemical Identifiers
| 888041-37-0 | |
| 386.086 | |
| BPGPBYGXGRDFQG-UHFFFAOYSA-N | |
| 2,7-dibromotriphenylene |
| C18H10Br2 | |
| MFCD22571695 | |
| 16122224 | |
| C1=CC=C2C(=C1)C3=C(C=CC(=C3)Br)C4=C2C=C(C=C4)Br |
Specifications
| 888041-37-0 | |
| Yellow | |
| MFCD22571695 | |
| BPGPBYGXGRDFQG-UHFFFAOYSA-N | |
| 2,7-dibromotriphenylene | |
| 16122224 | |
| ≥98.0% (GC) | |
| 2,7-Dibromotriphenylene |
| 232°C | |
| C18H10Br2 | |
| 200 mg | |
| C1=CC=C2C(=C1)C3=C(C=CC(=C3)Br)C4=C2C=C(C=C4)Br | |
| 386.086 | |
| 386.09 | |
| Crystalline Powder |
Safety and Handling
TSCA : No