missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,7-Dibromotriphenylene 98.0+%, TCI America™
Supplier: TCI America D48011G
Specifications
| 2,7-Dibromotriphenylene | |
| 232°C | |
| C18H10Br2 | |
| 1 g | |
| C1=CC=C2C(=C1)C3=C(C=CC(=C3)Br)C4=C2C=C(C=C4)Br | |
| 386.086 | |
| 386.09 | |
| Crystalline Powder |
| 888041-37-0 | |
| Yellow | |
| MFCD22571695 | |
| BPGPBYGXGRDFQG-UHFFFAOYSA-N | |
| 2,7-dibromotriphenylene | |
| 16122224 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 888041-37-0 | |
| 386.086 | |
| BPGPBYGXGRDFQG-UHFFFAOYSA-N | |
| 2,7-dibromotriphenylene |
| C18H10Br2 | |
| MFCD22571695 | |
| 16122224 | |
| C1=CC=C2C(=C1)C3=C(C=CC(=C3)Br)C4=C2C=C(C=C4)Br |
Safety and Handling
TSCA : No