missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,7-Dibromo-9-phenyl-9H-fluoren-9-ol 98.0+%, TCI America™
Supplier: TCI America D544825G
Specifications
| 2,7-Dibromo-9-phenyl-9H-fluoren-9-ol | |
| 165°C | |
| C19H12Br2O | |
| 25 g | |
| VGZUBRZNTWOSTO-UHFFFAOYSA-N | |
| 2,7-dibromo-9-phenylfluoren-9-ol | |
| 22117546 | |
| ≥98.0% (GC) |
| 132717-37-4 | |
| White-Yellow | |
| MFCD09953789 | |
| 2,7-Dibromo-9-hydroxy-9-phenyl-9H-fluorene | |
| C1=CC=C(C=C1)C2(C3=C(C=CC(=C3)Br)C4=C2C=C(C=C4)Br)O | |
| 416.112 | |
| 416.11 | |
| Crystalline Powder |
Chemical Identifiers
| 132717-37-4 | |
| 416.112 | |
| VGZUBRZNTWOSTO-UHFFFAOYSA-N | |
| 22117546 | |
| C1=CC=C(C=C1)C2(C3=C(C=CC(=C3)Br)C4=C2C=C(C=C4)Br)O |
| C19H12Br2O | |
| MFCD09953789 | |
| 2,7-Dibromo-9-hydroxy-9-phenyl-9H-fluorene | |
| 2,7-dibromo-9-phenylfluoren-9-ol |
Safety and Handling
TSCA : No