accessibility menu, dialog, popup

UserName

CAS RN 132717-37-4

IUPAC Name: 2,7-dibromo-9-phenyl-9H-fluoren-9-ol
Synonyms: 2,7-dibromo-9-phenylfluoren-9-ol
Molecular Weight (g/mol): 416.11
Molecular Formula: C19H12Br2O
InChi Key: VGZUBRZNTWOSTO-UHFFFAOYSA-N
SMILES: OC1(C2=C(C=CC(Br)=C2)C2=C1C=C(Br)C=C2)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results