missing translation for 'onlineSavingsMsg'
Learn More

2,7-Dibromo-9-phenyl-9H-fluoren-9-ol 98.0+%, TCI America™

Catalog No. D54485G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D54485G 5 g
D544825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D54485G Supplier TCI America Supplier No. D54485G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 132717-37-4
Molecular Formula C19H12Br2O
Molecular Weight (g/mol) 416.112
MDL Number MFCD09953789
InChI Key VGZUBRZNTWOSTO-UHFFFAOYSA-N
Synonym 2,7-Dibromo-9-hydroxy-9-phenyl-9H-fluorene
PubChem CID 22117546
IUPAC Name 2,7-dibromo-9-phenylfluoren-9-ol
SMILES C1=CC=C(C=C1)C2(C3=C(C=CC(=C3)Br)C4=C2C=C(C=C4)Br)O

Specifications

Melting Point 165°C
Color White-Yellow
Quantity 5 g
Formula Weight 416.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Dibromo-9-phenyl-9H-fluoren-9-ol