missing translation for 'onlineSavingsMsg'
Learn More

2,7-Dibromo-9,9-didecylfluorene 98.0+%, TCI America™

Catalog No. D45615G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45615G 5 g
D456125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45615G Supplier TCI America Supplier No. D45615G
Only null left

Chemical Identifiers

CAS 175922-78-8
Molecular Formula C33H48Br2
Molecular Weight (g/mol) 604.555
MDL Number MFCD00971288
InChI Key RLYANTSRYXOALQ-UHFFFAOYSA-N
PubChem CID 3661211
IUPAC Name 2,7-dibromo-9,9-didecylfluorene
SMILES CCCCCCCCCCC1(C2=C(C=CC(=C2)Br)C3=C1C=C(C=C3)Br)CCCCCCCCCC

Specifications

Melting Point 40°C
Color White-Yellow
Quantity 5 g
Formula Weight 604.56
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Dibromo-9,9-didecylfluorene