accessibility menu, dialog, popup

UserName

CH3BrBrCH3

CAS RN 175922-78-8

IUPAC Name: 2,7-dibromo-9,9-bis(decyl)-9H-fluorene
Synonyms: 2,7-dibromo-9,9-bis(decyl)fluorene
Molecular Weight (g/mol): 604.56
Molecular Formula: C33H48Br2
InChi Key: RLYANTSRYXOALQ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCC1(CCCCCCCCCC)C2=C(C=CC(Br)=C2)C2=C1C=C(Br)C=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)