missing translation for 'onlineSavingsMsg'
Learn More

2,7-Dibromo-9,9-bis(6-bromohexyl)fluorene 98.0+%, TCI America™

Catalog No. p-7123014
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35521G 1 g
B35525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B35521G Supplier TCI America Supplier No. B35521G

Chemical Identifiers

CAS 570414-33-4
Molecular Formula C25H30Br4
Molecular Weight (g/mol) 650.131
MDL Number MFCD09260450
InChI Key OJMAUBALNSWGDC-UHFFFAOYSA-N
PubChem CID 21867930
IUPAC Name 2,7-dibromo-9,9-bis(6-bromohexyl)fluorene
SMILES C1=CC2=C(C=C1Br)C(C3=C2C=CC(=C3)Br)(CCCCCCBr)CCCCCCBr

Specifications

Melting Point 72°C
Color White
Quantity 1 g
Formula Weight 650.13
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Dibromo-9,9-bis(6-bromohexyl)fluorene