missing translation for 'onlineSavingsMsg'
Learn More

2,7-Diaminofluorene 98.0+%, TCI America™

Catalog No. D00925G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D00925G 5 g
D00921G 1 g
D009225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D00925G Supplier TCI America Supplier No. D00925G

Chemical Identifiers

CAS 525-64-4
Molecular Formula C13H13ClN2
Molecular Weight (g/mol) 232.71
MDL Number MFCD00001128
InChI Key COOWDVCIUYIBFE-UHFFFAOYSA-N
Synonym 2,7-diaminofluorene, 2,7-fluorenediamine, 2,7-diamino-fluorene, 2,7-fluoroenediamine, fluorene-2,7-diamine, fluorene-2,7-diyldiamine, ccris 920, 2,7-diamino-fluoren, 2,7-diaminoflyorene, 2,7-diaminofluoren
PubChem CID 10679
IUPAC Name 7-amino-9H-fluoren-2-aminium chloride
SMILES [Cl-].NC1=CC=C2C(CC3=CC([NH3+])=CC=C23)=C1

Specifications

Melting Point 164°C
Quantity 5 g
Formula Weight 196.25
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Diaminofluorene
RTECSNumber LL6980000
TSCA No