accessibility menu, dialog, popup

UserName

ClNH2H3N

CAS RN 525-64-4

IUPAC Name: 7-amino-9H-fluoren-2-aminium chloride
Synonyms: 7-amino-9H-fluoren-2-aminium chloride
Molecular Weight (g/mol): 232.71
Molecular Formula: C13H13ClN2
InChi Key: COOWDVCIUYIBFE-UHFFFAOYSA-N
SMILES: [Cl-].NC1=CC=C2C(CC3=CC([NH3+])=CC=C23)=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)