missing translation for 'onlineSavingsMsg'
Learn More

2,7-Di-tert-butylfluorene 98.0+%, TCI America™

Catalog No. p-7128090
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40845G 5 g
D408425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D40845G Supplier TCI America Supplier No. D40845G
Only null left

Chemical Identifiers

CAS 58775-05-6
Molecular Formula C21H26
Molecular Weight (g/mol) 278.439
MDL Number MFCD03093998
InChI Key DFZYPLLGAQIQTD-UHFFFAOYSA-N
Synonym 2,7-di-tert-butylfluorene, 2,7-di-tert-butyl-9h-fluorene, 2,7-di-t-butylfluorene, 9h-fluorene, 2,7-bis 1,1-dimethylethyl, pubchem15066, 2,7-di-tert-butyl fluorene, 2,7-di-tert-butyl-fluorene, ksc491i2b
PubChem CID 4090082
IUPAC Name 2,7-ditert-butyl-9H-fluorene
SMILES CC(C)(C)C1=CC2=C(C=C1)C3=C(C2)C=C(C=C3)C(C)(C)C

Specifications

Melting Point 124°C
Color White-Yellow
Quantity 5 g
Formula Weight 278.44
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Di-tert-butylfluorene