missing translation for 'onlineSavingsMsg'
Learn More

2,7-Di-tert-butylfluorene, 97+%

Catalog No. p-7036942
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5543006 5 g
AAH5543014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5543006 Supplier Thermo Scientific Chemicals Supplier No. H5543006

CAS: 58775-05-6 | C21H26 | 278.439 g/mol

It is used as an organic light emitting diode (OLED) intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an organic light emitting diode (OLED) intermediate.

Solubility
Soluble in toluene, hot methanol.

Notes
Store at -20°C. Avoid heat and oxidizing agents.

Chemical Identifiers

CAS 58775-05-6
Molecular Formula C21H26
Molecular Weight (g/mol) 278.439
MDL Number MFCD03093998
InChI Key DFZYPLLGAQIQTD-UHFFFAOYSA-N
Synonym 2,7-di-tert-butylfluorene, 2,7-di-tert-butyl-9h-fluorene, 2,7-di-t-butylfluorene, 9h-fluorene, 2,7-bis 1,1-dimethylethyl, pubchem15066, 2,7-di-tert-butyl fluorene, 2,7-di-tert-butyl-fluorene, ksc491i2b
PubChem CID 4090082
IUPAC Name 2,7-ditert-butyl-9H-fluorene
SMILES CC(C)(C)C1=CC2=C(C=C1)C3=C(C2)C=C(C=C3)C(C)(C)C

Specifications

Melting Point 121°C to 124°C
Quantity 5 g
Solubility Information Soluble in toluene,hot methanol.
Formula Weight 278.43
Percent Purity 98%
Chemical Name or Material 2,7-Di-tert-butylfluorene
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only