missing translation for 'onlineSavingsMsg'
Learn More

2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9-didodecylfluorene 98.0+%, TCI America™

Catalog No. B46241G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B46241G Supplier TCI America Supplier No. B46241G
Only null left

Chemical Identifiers

CAS 749900-93-4
Molecular Formula C49H80B2O4
Molecular Weight (g/mol) 754.80
MDL Number MFCD28126781
InChI Key GFPPDTSBPULCFW-UHFFFAOYSA-N
Synonym 9,9-Didodecylfluorene-2,7-diboronic Acid Bis(pinacol) Ester
PubChem CID 58969154
IUPAC Name 2-[9,9-didodecyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-fluoren-2-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CCCCCCCCCCCCC1(CCCCCCCCCCCC)C2=C(C=CC(=C2)B2OC(C)(C)C(C)(C)O2)C2=C1C=C(C=C2)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 77°C
Color White-Yellow
Quantity 1 g
Formula Weight 754.80
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9-didodecylfluorene