missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dinitrobenzonitrile, 98+%

CAS: 35213-00-4 | C7H3N3O4 | 193.118 g/mol

$168.23 - $661.99

Chemical Identifiers

CAS 35213-00-4
Molecular Formula C7H3N3O4
Molecular Weight (g/mol) 193.118
MDL Number MFCD00041246
InChI Key ZYDGHQSJZAFMLU-UHFFFAOYSA-N
Synonym dinitrobenzonitrile, benzonitrile, 2,6-dinitro, 2,6-dinitro-benzonitrile, 2,6-dinitrobenzenecarbonitrile, 2,6-dinitrobenznitrile, pubchem21108, 2.6-dinitrobenzonitril, 2,6 dinitrobenzonitrile, 2,6-dini trobenzonitrile, acmc-1ad1y
PubChem CID 520718
IUPAC Name 2,6-dinitrobenzonitrile
SMILES C1=CC(=C(C(=C1)[N+](=O)[O-])C#N)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1262403
View Documents
Thermo Scientific Chemicals
L1262403
1 g
Each for $168.23
Only null left
 
AAL1262406
View Documents
Thermo Scientific Chemicals
L1262406
5 g
Each for $661.99
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

35213-00-4
193.118
ZYDGHQSJZAFMLU-UHFFFAOYSA-N
520718
C1=CC(=C(C(=C1)[N+](=O)[O-])C#N)[N+](=O)[O-]
C7H3N3O4
MFCD00041246
dinitrobenzonitrile, benzonitrile, 2,6-dinitro, 2,6-dinitro-benzonitrile, 2,6-dinitrobenzenecarbonitrile, 2,6-dinitrobenznitrile, pubchem21108, 2.6-dinitrobenzonitril, 2,6 dinitrobenzonitrile, 2,6-dini trobenzonitrile, acmc-1ad1y
2,6-dinitrobenzonitrile

Specifications

35213-00-4
C7H3N3O4
1 g
2215516
ZYDGHQSJZAFMLU-UHFFFAOYSA-N
2,6-dinitrobenzonitrile
520718
≥98%
146°C to 150°C
MFCD00041246
UN3439
dinitrobenzonitrile, benzonitrile, 2,6-dinitro, 2,6-dinitro-benzonitrile, 2,6-dinitrobenzenecarbonitrile, 2,6-dinitrobenznitrile, pubchem21108, 2.6-dinitrobenzonitril, 2,6 dinitrobenzonitrile, 2,6-dini trobenzonitrile, acmc-1ad1y
C1=CC(=C(C(=C1)[N+](=O)[O-])C#N)[N+](=O)[O-]
193.118
193.12
2,6-Dinitrobenzonitrile
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H373-H312-H332-H315-H319
Toxic if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c

H301-H312+H332-H315-H319-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only