CAS RN 35213-00-4
IUPAC Name:
2,6-dinitrobenzonitrile
Synonyms:
2,6-dinitrobenzonitrile
Molecular Weight (g/mol):
193.12
Molecular Formula:
C7H3N3O4
InChi Key:
ZYDGHQSJZAFMLU-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC=CC(=C1C#N)[N+]([O-])=O
1
–
1
of
1
results
1
Promotions Available
2,6-Dinitrobenzonitrile, 98+%
2,6-Dinitrobenzonitrile, 98+%
CAS: 35213-00-4 Molecular Formula: C7H3N3O4 Molecular Weight (g/mol): 193.118 MDL Number: MFCD00041246 InChI Key: ZYDGHQSJZAFMLU-UHFFFAOYSA-N Synonym: dinitrobenzonitrile,benzonitrile, 2,6-dinitro,2,6-dinitro-benzonitrile,2,6-dinitrobenzenecarbonitrile,2,6-dinitrobenznitrile,pubchem21108,2.6-dinitrobenzonitril,2,6 dinitrobenzonitrile,2,6-dini trobenzonitrile,acmc-1ad1y PubChem CID: 520718 IUPAC Name: 2,6-dinitrobenzonitrile SMILES: C1=CC(=C(C(=C1)[N+](=O)[O-])C#N)[N+](=O)[O-]