missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dinitrobenzonitrile, 98+%

Catalog No. AAL1262403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1262403 1 g
AAL1262406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1262403 Supplier Thermo Scientific Chemicals Supplier No. L1262403

CAS: 35213-00-4 | C7H3N3O4 | 193.118 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 35213-00-4
Molecular Formula C7H3N3O4
Molecular Weight (g/mol) 193.118
MDL Number MFCD00041246
InChI Key ZYDGHQSJZAFMLU-UHFFFAOYSA-N
Synonym dinitrobenzonitrile, benzonitrile, 2,6-dinitro, 2,6-dinitro-benzonitrile, 2,6-dinitrobenzenecarbonitrile, 2,6-dinitrobenznitrile, pubchem21108, 2.6-dinitrobenzonitril, 2,6 dinitrobenzonitrile, 2,6-dini trobenzonitrile, acmc-1ad1y
PubChem CID 520718
IUPAC Name 2,6-dinitrobenzonitrile
SMILES C1=CC(=C(C(=C1)[N+](=O)[O-])C#N)[N+](=O)[O-]

Specifications

Melting Point 146°C to 150°C
Quantity 1 g
UN Number UN3439
Beilstein 2215516
Formula Weight 193.12
Percent Purity ≥98%
Chemical Name or Material 2,6-Dinitrobenzonitrile
Hazard Category H301-H312+H332-H315-H319-H373
Hazard Statement GHS H Statement
H301-H373-H312-H332-H315-H319
Toxic if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only