missing translation for 'onlineSavingsMsg'
Learn More

(2,6-Dimethylphenyl)thiourea 98.0+%, TCI America™

Catalog No. D49585G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49585G 5 g
D495825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49585G Supplier TCI America Supplier No. D49585G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6396-76-5
Molecular Formula C9H12N2S
Molecular Weight (g/mol) 180.27
MDL Number MFCD00041165
InChI Key ASNKJUONFPQYPC-UHFFFAOYSA-N
Synonym 1-2,6-dimethylphenyl thiourea, 1-2,6-dimethylphenyl-2-thiourea, 2,6-dimethylphenyl thiourea, n-2,6-dimethylphenyl thiourea, 1-2,6-xylyl thiourea, thiourea, 2,6-dimethylphenyl, amino 2,6-dimethylphenyl amino methane-1-thione, acmc-20apd5, 1-2,6-xylyl-2-thiourea, 2,6-dimethyl-phenyl-thiourea
PubChem CID 853911
IUPAC Name (2,6-dimethylphenyl)thiourea
SMILES CC1=CC=CC(C)=C1NC(N)=S

Specifications

Melting Point 192°C
Color White-Yellow
Quantity 5 g
UN Number 2811
Formula Weight 180.27
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material (2,6-Dimethylphenyl)thiourea