accessibility menu, dialog, popup

UserName

CAS RN 6396-76-5

IUPAC Name: (2,6-dimethylphenyl)thiourea
Synonyms: 2,6-dimethylphenylthiourea
Molecular Weight (g/mol): 180.27
Molecular Formula: C9H12N2S
InChi Key: ASNKJUONFPQYPC-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C)=C1NC(N)=S
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results