missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylphenoxyacetic Acid 98.0+%, TCI America™

Catalog No. p-7128236
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43451G 1 g
D43455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43451G Supplier TCI America Supplier No. D43451G

Chemical Identifiers

CAS 13335-71-2
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00156912
InChI Key MLBCURLNKYKBEQ-UHFFFAOYSA-N
PubChem CID 101369
IUPAC Name 2-(2,6-dimethylphenoxy)acetic acid
SMILES CC1=C(C(=CC=C1)C)OCC(=O)O

Specifications

Melting Point 139°C
Quantity 1 g
Formula Weight 180.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dimethylphenoxyacetic Acid