accessibility menu, dialog, popup

UserName

CH3H3COHOO

CAS RN 13335-71-2

IUPAC Name: 2-(2,6-dimethylphenoxy)acetic acid
Synonyms: 2,6-dimethylphenoxyacetic acid
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: MLBCURLNKYKBEQ-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C)=C1OCC(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results