missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylnitrobenzene 99.0+%, TCI America™

Catalog No. D027325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D027325G 25 g
D0273500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D027325G Supplier TCI America Supplier No. D027325G

Chemical Identifiers

CAS 81-20-9
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00007181
InChI Key HDFQKJQEWGVKCQ-UHFFFAOYSA-N
Synonym 2-nitro-m-xylene, 2,6-dimethylnitrobenzene, benzene, 1,3-dimethyl-2-nitro, 2,6-dimethyl-1-nitrobenzene, m-xylene, 2-nitro, ccris 3119, 1,3-dimethyl-2-nitro-benzene, dsstox_cid_5134, dsstox_rid_77678, dsstox_gsid_25134
PubChem CID 6672
IUPAC Name 1,3-dimethyl-2-nitrobenzene
SMILES CC1=C(C(=CC=C1)C)[N+](=O)[O-]

Specifications

Color Yellow
Boiling Point 222°C
Quantity 25 g
UN Number 1665
Formula Weight 151.17
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,6-Dimethylnitrobenzene
EINECSNumber (3)-0399
RTECSNumber ZE4686000
TSCA Yes