missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol 98.0+%, TCI America™

Catalog No. D28531G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28531G 1 g
D28535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D28531G Supplier TCI America Supplier No. D28531G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 269410-25-5
Molecular Formula C14H21BO3
Molecular Weight (g/mol) 248.13
MDL Number MFCD02093724
InChI Key TYCKOBOJYNRIBO-UHFFFAOYSA-N
Synonym 2,6-dimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 4-hydroxy-3,5-dimethylphenylboronic acid pinacol ester, 3,5-dimethyl-4-hydroxyphenylboronic acid pinacol ester, 3,5-dimethyl-4-hydroxybenzeneboronic acid, pinacol ester, 4-hydroxy-3,5-dimethylphenylboronic acid, pinacol ester, 2,6-dimethyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 2,6-dimethyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenol, 2,6-dimethyl-4-4,4-5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 2-4-hydroxy-3,5-dimethylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, phenol, 2,6-dimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
PubChem CID 2734649
IUPAC Name 2,6-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
SMILES CC1=CC(=CC(C)=C1O)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 108°C
Quantity 1 g
Formula Weight 248.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol