accessibility menu, dialog, popup

UserName

CAS RN 269410-25-5

IUPAC Name: 2,6-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Synonyms: 2,6-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Molecular Weight (g/mol): 248.13
Molecular Formula: C14H21BO3
InChi Key: TYCKOBOJYNRIBO-UHFFFAOYSA-N
SMILES: CC1=CC(=CC(C)=C1O)B1OC(C)(C)C(C)(C)O1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results