missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethoxy-3-nitrobenzoic acid, 97%

Catalog No. AAL0436614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0436614 25 g
AAL0436606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0436614 Supplier Thermo Scientific Chemicals Supplier No. L0436614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 55776-17-5 | C9H9NO6 | 227.172 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 55776-17-5
Molecular Formula C9H9NO6
Molecular Weight (g/mol) 227.172
MDL Number MFCD00017324
InChI Key YIKBQFPTPKEFSM-UHFFFAOYSA-N
Synonym 2,6-dimethoxy-3-nitro-benzoic acid, acmc-20ao8d, benzoic acid, 2,6-dimethoxy-3-nitro
PubChem CID 3085094
IUPAC Name 2,6-dimethoxy-3-nitrobenzoic acid
SMILES COC1=C(C(=C(C=C1)[N+](=O)[O-])OC)C(=O)O

Specifications

Melting Point 130°C to 132°C
Quantity 25 g
Beilstein 2657939
Formula Weight 227.17
Percent Purity 97%
Chemical Name or Material 2,6-Dimethoxy-3-nitrobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 259-814-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only