CAS RN 55776-17-5
IUPAC Name:
2,6-dimethoxy-3-nitrobenzoic acid
Synonyms:
2,6-dimethoxy-3-nitrobenzoic acid
Molecular Weight (g/mol):
227.17
Molecular Formula:
C9H9NO6
InChi Key:
YIKBQFPTPKEFSM-UHFFFAOYSA-N
SMILES:
COC1=CC=C(C(OC)=C1C(O)=O)[N+]([O-])=O
1
–
1
of
1
results
1
Promotions Available
2,6-Dimethoxy-3-nitrobenzoic acid, 97%
2,6-Dimethoxy-3-nitrobenzoic acid, 97%
CAS: 55776-17-5 Molecular Formula: C9H9NO6 Molecular Weight (g/mol): 227.172 MDL Number: MFCD00017324 InChI Key: YIKBQFPTPKEFSM-UHFFFAOYSA-N Synonym: 2,6-dimethoxy-3-nitro-benzoic acid,acmc-20ao8d,benzoic acid, 2,6-dimethoxy-3-nitro PubChem CID: 3085094 IUPAC Name: 2,6-dimethoxy-3-nitrobenzoic acid SMILES: COC1=C(C(=C(C=C1)[N+](=O)[O-])OC)C(=O)O