missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diisopropylaniline 90.0+%, TCI America™

Catalog No. D175525ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D175525ML 25 mL
D1755100ML 100 mL
D1755500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D175525ML Supplier TCI America Supplier No. D175525ML

Chemical Identifiers

CAS 24544-04-5
Molecular Formula C12H19N
Molecular Weight (g/mol) 177.291
MDL Number MFCD00008887
InChI Key WKBALTUBRZPIPZ-UHFFFAOYSA-N
Synonym 2,6-diisopropylaniline, benzenamine, 2,6-bis 1-methylethyl, 2,6-diisopropyl aniline, aniline, 2,6-diisopropyl, diisopropylaniline, 2,6-diisopropylphenylamine, 2,6-bis propan-2-yl aniline, 2,6-dilsopropylaniline, 2,6-di propan-2-yl aniline, 2,6-bis 1-methylethyl aniline
PubChem CID 32484
IUPAC Name 2,6-di(propan-2-yl)aniline
SMILES CC(C)C1=C(C(=CC=C1)C(C)C)N

Specifications

Melting Point -45°C
Boiling Point 122°C
Quantity 25 mL
Formula Weight 177.29
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,6-Diisopropylaniline
EINECSNumber (3)-0129
RTECSNumber BX4025000
TSCA Yes