accessibility menu, dialog, popup

UserName

CAS RN 24544-04-5

IUPAC Name: 2,6-bis(propan-2-yl)aniline
Synonyms: 2,6-diisopropylaniline
Molecular Weight (g/mol): 177.29
Molecular Formula: C12H19N
InChi Key: WKBALTUBRZPIPZ-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=CC(C(C)C)=C1N
Boiling Point 
  • (3)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (5)