missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diisopropylaniline, 90+%

Catalog No. AC169411000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC169411000 100 g Glass bottle
AC169415000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC169411000 Supplier Thermo Scientific Chemicals Supplier No. 169411000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 24544-04-5 | C12H19N | 177.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 24544-04-5
Molecular Formula C12H19N
Molecular Weight (g/mol) 177.29
MDL Number MFCD00008887
InChI Key WKBALTUBRZPIPZ-UHFFFAOYSA-N
Synonym 2,6-diisopropylaniline, benzenamine, 2,6-bis 1-methylethyl, 2,6-diisopropyl aniline, aniline, 2,6-diisopropyl, diisopropylaniline, 2,6-diisopropylphenylamine, 2,6-bis propan-2-yl aniline, 2,6-dilsopropylaniline, 2,6-di propan-2-yl aniline, 2,6-bis 1-methylethyl aniline
PubChem CID 32484
IUPAC Name 2,6-di(propan-2-yl)aniline
SMILES CC(C)C1=C(C(=CC=C1)C(C)C)N

Specifications

CAS Min % 90.0
CAS Max % 100.0
Melting Point -45.0°C
Density 0.9400g/mL
Boiling Point 257.0°C
Flash Point 123°C
Infrared Spectrum Authentic
Assay Percent Range 90% min. (GC)
Packaging Glass bottle
Refractive Index 1.5320 to 1.5340
Linear Formula [(CH3)2CH]2C6H3NH2
Quantity 100 g
Beilstein 12, 168
Specific Gravity 0.94
Solubility Information Solubility in water: <0.20g/L. Other solubilities: miscible with ethanol and toluene
Formula Weight 177.29
Percent Purity 90+%
Physical Form Liquid
Chemical Name or Material 2, 6-Diisopropylaniline, 92%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 246-305-4
RTECSNumber BX4025000
TSCA TSCA

RUO – Research Use Only