missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diisopropyl-N,N-dimethylaniline, 96%

Catalog No. AAH2727503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2727503 1 g
AAH2727506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2727503 Supplier Thermo Scientific Chemicals Supplier No. H2727503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2909-77-5 | C14H23N | 205.345 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2909-77-5
Molecular Formula C14H23N
Molecular Weight (g/mol) 205.345
MDL Number MFCD00144978
InChI Key ALXIOUGHHXXLKX-UHFFFAOYSA-N
Synonym 2,6-diisopropyl-n,n-dimethylaniline, n,n-dimethyl-2,6-diisopropylaniline, benzenamine, n,n-dimethyl-2,6-bis 1-methylethyl, 2,6-diisopropyl-n,n-dimethyl-aniline, acmc-20ao8g, 2,6-bis methylethyl phenyl dimethylamine, n,n-dimethyl-2,6-di propan-2-yl aniline, benzenamine,n,n-dimethyl-2,6-bis 1-methylethyl
PubChem CID 76198
IUPAC Name N,N-dimethyl-2,6-di(propan-2-yl)aniline
SMILES CC(C)C1=C(C(=CC=C1)C(C)C)N(C)C

Specifications

Density 0.873
Boiling Point 86°C (2.5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5005
Quantity 1 g
UN Number UN2810
Formula Weight 205.34
Percent Purity 96%
Chemical Name or Material 2,6-Diisopropyl-N,N-dimethylaniline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only