missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dihydroxyanthracene 95.0+%, TCI America™
$210.63 - $631.92
Chemical Identifiers
| CAS | 101488-73-7 |
|---|---|
| Molecular Formula | C14H10O2 |
| Molecular Weight (g/mol) | 210.232 |
| InChI Key | JBBHFHMVEOHFRE-UHFFFAOYSA-N |
| Synonym | 2,6-Anthracenediol |
| PubChem CID | 13466096 |
| IUPAC Name | anthracene-2,6-diol |
| SMILES | C1=CC(=CC2=CC3=C(C=C21)C=C(C=C3)O)O |
Chemical Identifiers
| 101488-73-7 | |
| 210.232 | |
| 2,6-Anthracenediol | |
| anthracene-2,6-diol |
| C14H10O2 | |
| JBBHFHMVEOHFRE-UHFFFAOYSA-N | |
| 13466096 | |
| C1=CC(=CC2=CC3=C(C=C21)C=C(C=C3)O)O |
Specifications
| 101488-73-7 | |
| C14H10O2 | |
| 2,6-Anthracenediol | |
| C1=CC(=CC2=CC3=C(C=C21)C=C(C=C3)O)O | |
| 210.232 | |
| 210.23 | |
| Crystalline Powder |
| 300°C | |
| 200 mg | |
| JBBHFHMVEOHFRE-UHFFFAOYSA-N | |
| anthracene-2,6-diol | |
| 13466096 | |
| ≥95.0% (GC) | |
| 2,6-Dihydroxyanthracene |
Safety and Handling
TSCA : No