accessibility menu, dialog, popup

UserName

CAS RN 101488-73-7

OHHO

CAS RN 101488-73-7

IUPAC Name: anthracene-2,6-diol
Synonyms: anthracene-2,6-diol
Molecular Weight (g/mol): 210.23
Molecular Formula: C14H10O2
InChi Key: JBBHFHMVEOHFRE-UHFFFAOYSA-N
SMILES: OC1=CC2=CC3=CC=C(O)C=C3C=C2C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results